Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033467.3 | NP_258428.2 | 20 | 2157 | ||
XM_011542122.2 | XP_011540424.1 | 13 | 1260 | |||
XM_017002310.1 | XP_016857799.1 | 21 | 2423 | |||
XM_017002311.1 | XP_016857800.1 | 20 | 2103 | |||
XM_017002312.1 | XP_016857801.1 | 19 | 2075 | |||
XM_017002313.1 | XP_016857802.1 | 20 | 2157 | |||
XM_017002314.1 | XP_016857803.1 | 19 | 2028 | |||
XM_017002315.1 | XP_016857804.1 | 20 | 2033 | |||
GenBank mRNA | AF336981.1 | - | 19 | 1980 | ||
AK093058.1 | - | 20 | 2105 | |||
BC101027.2 | - | 19 | 1953 | 40007698 | ||
BC101028.1 | - | 15 | 1482 | 40007699 | ||
BC101029.2 | - | 19 | 1982 | 40007702 | ||
BC101030.1 | - | 19 | 1953 | 40007706 |