Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030907.3 | NP_112169.2 | 3 | 777 | ||
XM_011542126.2 | XP_011540428.1 | 3 | 1058 | |||
GenBank mRNA | AL559024.3 | - | 3 | 707 | ||
BC002946.2 | - | 3 | 564 | 3955163 | ||
BC008702.1 | - | 3 | 564 | 2966866 |