Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024520.2 | NP_078796.2 | 5 | 1054 | ||
GenBank mRNA | AK026208.1 | - | 5 | 1054 | ||
AL832964.1 | - | 4 | 373 | |||
BC017959.1 | - | 4 | 738 | 4607945 | ||
BQ448209.1 | - | 2 | 468 |