Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024528.3 | NP_078804.2 | 8 | 1194 | ||
XM_017029842.1 | XP_016885331.1 | 8 | 731 | |||
GenBank mRNA | AK026279.1 | - | 8 | 1194 | ||
AK094446.1 | - | 6 | 1955 | |||
AL832118.1 | - | 10 | 1135 | |||
AY388958.1 | - | 8 | 1194 | |||
BC000940.2 | - | 8 | 1076 | |||
BC015354.1 | - | 8 | 1058 | 3450847 | ||
BC032442.1 | - | 8 | 1052 | 5139142 |