Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077416.2 | NP_001070884.2 | 4 | 791 | ||
NM_001077418.2 | NP_001070886.1 | 5 | 694 | |||
NR_074083.1 | 5 | 817 | ||||
GenBank mRNA | AK025820.1 | - | 5 | 666 | ||
AK096650.1 | - | 4 | 743 | |||
AK290483.1 | - | 5 | 680 | |||
AK298994.1 | - | 2 | 388 | |||
AK307263.1 | - | 5 | 783 | |||
AY358612.1 | - | 5 | 690 | |||
BC010609.1 | - | 5 | 648 | 4214587 | ||
BC016401.1 | - | 4 | 326 | |||
BC063677.1 | - | 5 | 799 |