Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077416.2 | NP_001070884.2 | 5 | 892 | ||
NM_001077418.2 | NP_001070886.1 | 6 | 795 | |||
NR_074083.1 | 6 | 918 | ||||
GenBank mRNA | AK025820.1 | - | 6 | 767 | ||
AK057689.1 | - | 1 | 644 | |||
AK096650.1 | - | 5 | 844 | |||
AK290483.1 | - | 6 | 781 | |||
AK298994.1 | - | 3 | 489 | |||
AY358612.1 | - | 6 | 791 | |||
BC010609.1 | - | 6 | 749 | 4214587 | ||
BC016401.1 | - | 5 | 427 | |||
BC063677.1 | - | 6 | 900 |