Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077416.2 | NP_001070884.2 | 5, 6 | 945 | ||
NM_001077418.2 | NP_001070886.1 | 6, 7 | 848 | |||
NR_074083.1 | 6, 7 | 971 | ||||
GenBank mRNA | AK025820.1 | - | 6, 7 | 820 | ||
AK057689.1 | - | 1, 2 | 697 | |||
AK096650.1 | - | 5, 6 | 897 | |||
AK290483.1 | - | 6, 7 | 834 | |||
AK298994.1 | - | 3, 4 | 542 | |||
AY358612.1 | - | 6, 7 | 844 | |||
BC010609.1 | - | 6, 7 | 802 | 4214587 | ||
BC016401.1 | - | 5, 6 | 480 | |||
BC063677.1 | - | 6, 7 | 953 |