Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024537.3 | NP_078813.1 | 7, 8 | 877 | ||
XM_006719953.2 | XP_006720016.1 | 7, 8 | 957 | |||
XM_011521116.1 | XP_011519418.1 | 8, 9 | 719 | |||
XM_011521117.2 | XP_011519419.1 | 7, 8 | 813 | |||
XM_011521118.2 | XP_011519420.1 | 7, 8 | 813 | |||
XM_017020741.1 | XP_016876230.1 | 6, 7 | 605 | |||
XM_017020742.1 | XP_016876231.1 | 7, 8 | 813 | |||
GenBank mRNA | AB062436.1 | - | 7, 8 | 794 | ||
AK022180.1 | - | 7, 8 | 813 | |||
AK301907.1 | - | 6, 7 | 982 | |||
BC007220.1 | - | 7, 8 | 800 | 2822557 |