Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TANGO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024562.1 | NP_078838.1 | 10 | 1787 | ||
XM_011523327.2 | XP_011521629.1 | 10 | 1846 | |||
GenBank mRNA | AB051533.1 | - | 10 | 1787 | ||
AK022750.1 | - | 4 | 451 | |||
AK095333.1 | - | 9 | 1333 |