Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024565.6 | NP_078841.3 | 3 | 553 | ||
XM_006714917.3 | XP_006714980.1 | 2 | 988 | |||
XM_011534646.2 | XP_011532948.1 | 2 | 291 | |||
XM_017009846.1 | XP_016865335.1 | 3 | 636 | |||
XM_017009847.1 | XP_016865336.1 | 3 | 635 | |||
GenBank mRNA | BC035871.1 | - | 3 | 519 | 5245637 |