Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142683.2 | NP_001136155.1 | 2 | 1247 | ||
NM_024584.4 | NP_078860.2 | 2 | 846 | |||
XM_005264560.3 | XP_005264617.1 | 2 | 1366 | |||
GenBank mRNA | AK023708.1 | - | 1 | 342 | ||
AK124337.1 | - | 1 | 1830 | |||
AK125354.1 | - | 2 | 827 | |||
BC066936.1 | - | 2 | 799 | 4375206 | ||
BC066969.1 | - | 2 | 1247 | 5272989 |