Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005498.3 | NP_001005498.2 | 8 | 1207 | ||
NM_024599.5 | NP_078875.4 | 9 | 1369 | |||
XM_005257669.3 | XP_005257726.1 | 9 | 1730 | |||
XM_005257670.1 | XP_005257727.1 | 9 | 1239 | |||
XM_011525249.2 | XP_011523551.1 | 9 | 2686 | |||
XM_011525250.2 | XP_011523552.1 | 10 | 1396 | |||
XM_011525251.2 | XP_011523553.1 | 9 | 1213 | |||
XM_017025079.1 | XP_016880568.1 | 9 | 1643 | |||
GenBank mRNA | AK025994.1 | - | 4 | 393 | ||
AK057882.1 | - | 6 | 1063 | |||
AK074034.1 | - | 7 | 1674 | |||
AK292135.1 | - | 9 | 1362 | |||
AK292166.1 | - | 8 | 1162 | |||
AK303434.1 | - | 7 | 1023 | |||
AL833680.1 | - | 3 | 262 | |||
AY500247.1 | - | 8 | 1058 | |||
BC016034.1 | - | 9 | 1349 | 3910543 | ||
BC035829.1 | - | 4 | 455 |