Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302082.1 | NP_001289011.1 | 4 | 1023 | ||
NM_024603.3 | NP_078879.2 | 4 | 941 | |||
XM_011542141.2 | XP_011540443.1 | 4 | 3441 | |||
XM_011542142.2 | XP_011540444.1 | 3 | 889 | |||
XM_017002331.1 | XP_016857820.1 | 5 | 974 | |||
XM_017002332.1 | XP_016857821.1 | 5 | 851 | |||
XM_017002333.1 | XP_016857822.1 | 5 | 2639 | |||
XM_017002334.1 | XP_016857823.1 | 5 | 1070 | |||
GenBank mRNA | AK021650.1 | - | 4 | 885 | ||
AK055449.1 | - | 2 | 1574 | |||
BC007932.2 | - | 4 | 893 | 4121760 | ||
BC011804.2 | - | 4 | 707 | 4336159 | ||
BQ006447.1 | - | 3 | 491 | |||
KU178776.1 | - | 4 | 803 | |||
KU178777.1 | - | 4 | 885 | |||
KU178778.1 | - | 3 | 669 |