Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018089.2 | NP_001018099.1 | 11, 12 | 2389 | ||
NM_024611.5 | NP_078887.2 | 11, 12 | 2549 | |||
XM_005254661.1 | XP_005254718.1 | 12, 13 | 2548 | |||
XM_005254662.1 | XP_005254719.1 | 12, 13 | 2411 | |||
XM_011522009.1 | XP_011520311.1 | 11, 12 | 2423 | |||
XM_011522010.1 | XP_011520312.1 | 11, 12 | 2292 | |||
XM_011522011.2 | XP_011520313.1 | 10, 11 | 2206 | |||
XM_017022569.1 | XP_016878058.1 | 12, 13 | 2585 | |||
GenBank mRNA | AF502591.1 | - | 11, 12 | 2492 | ||
AK021958.1 | - | 2, 3 | 543 | |||
AK055752.1 | - | 11, 12 | 2492 | |||
AK095389.1 | - | 11, 12 | 2373 | |||
AK127892.1 | - | 4, 5 | 1374 | |||
AL832046.1 | - | 11, 12 | 2516 | |||
BC013684.1 | - | 2, 3 | 544 | |||
BC071685.1 | - | 2, 3 | 438 | |||
BC107427.1 | - | 2, 3 | 522 | |||
BC140894.1 | - | 11, 12 | 2468 | 9021762 | ||
BX647873.1 | - | 11, 12 | 2375 | |||
CR749852.1 | - | 11, 12 | 2342 |