Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024629.3 | NP_078905.2 | 12 | 1197 | ||
NR_104593.1 | 11 | 1135 | ||||
XM_005263218.4 | XP_005263275.2 | 12 | 1232 | |||
GenBank mRNA | AF469667.1 | - | 12 | 1200 | ||
AF516710.1 | - | 13 | 1614 | |||
AK027121.1 | - | 10 | 1007 | |||
AK292297.1 | - | 12 | 1143 | |||
BC031520.1 | - | 7 | 545 | |||
BC067802.1 | - | 11 | 1075 | |||
BC107744.1 | - | 7 | 642 | |||
BC131556.1 | - | 12 | 1197 | 40108113 | ||
BC141854.1 | - | 11 | 1133 | 8860370 | ||
BG031878.1 | - | 1 | 39 | |||
CR457376.1 | - | 5 | 401 |