Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024630.2 | NP_078906.2 | 9 | 1796 | ||
NM_153746.1 | NP_714968.1 | 9 | 1751 | |||
XM_005267146.3 | XP_005267203.1 | 8 | 1167 | |||
XM_011536120.2 | XP_011534422.1 | 9 | 2377 | |||
XM_011536123.2 | XP_011534425.1 | 9 | 1188 | |||
XM_017011307.1 | XP_016866796.1 | 9 | 1481 | |||
XM_017011309.1 | XP_016866798.1 | 9 | 1436 | |||
XM_017011311.1 | XP_016866800.1 | 8 | 1343 | |||
XM_017011312.1 | XP_016866801.1 | 9 | 1496 | |||
XM_017011313.1 | XP_016866802.1 | 10 | 4193 | |||
GenBank mRNA | AF542388.1 | - | 9 | 1796 | ||
AF542389.1 | - | 9 | 1751 | |||
AJ420552.1 | - | 1 | 28 | |||
AK024637.1 | - | 5 | 605 | |||
AK092380.1 | - | 5 | 2307 | |||
AK124573.1 | - | 6 | 4163 | |||
AK128773.1 | - | 5 | 3070 | |||
BC029881.1 | - | 8 | 910 | |||
BC056399.1 | - | 3 | 438 | |||
BC125072.1 | - | 10 | 1313 | |||
BC125073.1 | - | 10 | 1313 |