Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205315.1 | NP_001192244.1 | 6 | 1486 | ||
NM_001205316.1 | NP_001192245.1 | 4 | 859 | |||
NM_024636.3 | NP_078912.2 | 5 | 1387 | |||
GenBank mRNA | AF423422.1 | - | 6 | 1369 | ||
AF423423.1 | - | 6 | 1370 | |||
AK026806.1 | - | 5 | 1368 | |||
AL832955.1 | - | 3 | 478 | |||
AL833044.1 | - | 3 | 292 | |||
AL833671.1 | - | 2 | 208 | |||
BC020600.1 | - | 4 | 834 | 4616389 |