Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024640.3 | NP_078916.3 | 4 | 659 | ||
GenBank mRNA | AF250397.1 | - | 4 | 478 | ||
AK027129.1 | - | 3 | 263 | |||
AK129676.1 | - | 1 | 1181 | |||
AL524812.3 | - | 4 | 693 | |||
AY172561.1 | - | 4 | 651 | |||
AY286019.1 | - | 4 | 640 | |||
AY286020.1 | - | 4 | 640 | |||
BC008984.1 | - | 4 | 686 | |||
BC067857.1 | - | 4 | 644 | |||
BC068057.1 | - | 4 | 621 | |||
BC080186.1 | - | 4 | 661 | 6147134 |