Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024652.4 | NP_078928.3 | 14 | 2154 | ||
XM_011522012.2 | XP_011520314.1 | 16 | 2428 | |||
XM_011522013.2 | XP_011520315.1 | 14 | 2137 | |||
XM_011522014.2 | XP_011520316.1 | 14 | 2352 | |||
XM_011522015.2 | XP_011520317.1 | 9 | 2947 | |||
XM_011522016.2 | XP_011520318.1 | 12 | 1629 | |||
XM_011522019.2 | XP_011520321.1 | 14 | 2352 | |||
XM_017022570.1 | XP_016878059.1 | 4 | 401 | |||
GenBank mRNA | BC144705.1 | - | 13 | 1825 | ||
BC171802.1 | - | 13 | 1825 | |||
DQ013130.1 | - | 13 | 1792 |