Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024652.4 | NP_078928.3 | 31 | 5252 | ||
XM_005254979.3 | XP_005255036.1 | 17 | 2910 | |||
XM_011522012.2 | XP_011520314.1 | 33 | 5526 | |||
XM_011522013.2 | XP_011520315.1 | 31 | 5235 | |||
XM_011522014.2 | XP_011520316.1 | 31 | 5450 | |||
XM_011522015.2 | XP_011520317.1 | 26 | 6045 | |||
XM_011522016.2 | XP_011520318.1 | 29 | 4727 | |||
XM_011522017.2 | XP_011520319.1 | 17 | 2917 | |||
XM_011522018.2 | XP_011520320.1 | 17 | 2912 | |||
XM_017022570.1 | XP_016878059.1 | 21 | 3499 | |||
GenBank mRNA | AB058693.1 | - | 17 | 2957 | ||
AK054662.1 | - | 5 | 700 | |||
AK058028.1 | - | 11 | 1800 | |||
AK131075.1 | - | 12 | 2185 | |||
BC005408.1 | - | 6 | 923 | |||
BC020379.1 | - | 6 | 923 | |||
BC066655.1 | - | 2 | 165 | |||
BC144705.1 | - | 29 | 4640 | |||
BC146769.1 | - | 17 | 2957 | |||
BC171802.1 | - | 30 | 5119 | |||
DQ013130.1 | - | 30 | 4890 |