Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
siRNA ID
HSS188136
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
7
876
NM_001164629.3
NP_001158101.1
7
861
NM_001284233.1
NP_001271162.1
5
685
NM_001284234.1
NP_001271163.1
5
576
NM_001284235.1
NP_001271164.1
7
843
NM_001284238.1
NP_001271167.1
9
1075
NM_024659.4
NP_078935.2
7
843
XM_005263774.2
XP_005263831.1
8
960
XM_005263782.2
XP_005263839.1
8
976
XM_005263784.2
XP_005263841.1
6
835
XM_011511843.2
XP_011510145.1
8
1009
XM_011511855.2
XP_011510157.1
6
953
XM_011511856.2
XP_011510158.1
6
735
XM_017004917.1
XP_016860406.1
12
1631
XM_017004918.1
XP_016860407.1
10
1223
XM_017004919.1
XP_016860408.1
8
1109
XM_017004920.1
XP_016860409.1
10
4265
XM_017004921.1
XP_016860410.1
9
1058
XM_017004922.1
XP_016860411.1
9
1235
XM_017004923.1
XP_016860412.1
8
1023
XM_017004924.1
XP_016860413.1
10
1219
XM_017004925.1
XP_016860414.1
7
881
XM_017004926.1
XP_016860415.1
5
812
XM_017004927.1
XP_016860416.1
7
843
XM_017004928.1
XP_016860417.1
9
1076
XM_017004929.1
XP_016860418.1
7
813
XM_017004930.1
XP_016860419.1
7
1464
XM_017004931.1
XP_016860420.1
7
877
XM_017004932.1
XP_016860421.1
7
847
XM_017004933.1
XP_016860422.1
7
829
XM_017004934.1
XP_016860423.1
8
4036
XM_017004935.1
XP_016860424.1
9
1906
XM_017004936.1
XP_016860425.1
8
1083
XM_017004937.1
XP_016860426.1
6
964
XM_017004938.1
XP_016860427.1
6
962
XM_017004939.1
XP_016860428.1
8
1012
XM_017004940.1
XP_016860429.1
5
813
XM_017004941.1
XP_016860430.1
5
812
XM_017004942.1
XP_016860431.1
7
859
XM_017004943.1
XP_016860432.1
7
1525
XM_017004944.1
XP_016860433.1
7
858
XM_017004945.1
XP_016860434.1
9
1076
XM_017004946.1
XP_016860435.1
7
813
XM_017004947.1
XP_016860436.1
7
1496
XM_017004948.1
XP_016860437.1
8
4036
GenBank mRNA
AB201269.1
-
4
597
AB597184.1
-
4
597
AK021815.1
-
1
52
AK091684.1
-
5
689
AK291357.1
-
7
791
AY281366.1
-
7
876
BC017741.2
-
8
883
4390876
BC061699.1
-
7
783
4608518
BX647773.1
-
9
1014
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline