Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN28A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024674.5 | NP_078950.1 | 3 | 513 | ||
XM_011542148.1 | XP_011540450.1 | 3 | 503 | |||
GenBank mRNA | AF521099.1 | - | 3 | 464 | ||
AK022519.1 | - | 3 | 464 | |||
BC028566.2 | - | 3 | 503 | 3841184 | ||
CN349325.1 | - | 3 | 515 |