Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024694.3 | NP_078970.3 | 22 | 2807 | ||
XM_006715566.3 | XP_006715629.1 | 22 | 2842 | |||
XM_011536125.2 | XP_011534427.1 | 22 | 2793 | |||
XM_011536126.1 | XP_011534428.1 | 22 | 2842 | |||
XM_011536127.2 | XP_011534429.1 | 22 | 2842 | |||
XM_011536128.2 | XP_011534430.1 | 22 | 2842 | |||
XM_011536130.2 | XP_011534432.1 | 23 | 2741 | |||
XM_011536131.2 | XP_011534433.1 | 16 | 2015 | |||
XM_011536132.2 | XP_011534434.1 | 14 | 1603 | |||
XM_011536134.2 | XP_011534436.1 | 18 | 4296 | |||
XM_011536135.2 | XP_011534437.1 | 4 | 484 | |||
XM_017011314.1 | XP_016866803.1 | 22 | 2842 | |||
XM_017011315.1 | XP_016866804.1 | 22 | 2842 | |||
GenBank mRNA | AL832192.1 | - | 18 | 4298 | ||
DN831198.1 | - | 2 | 123 |