Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024713.2 | NP_078989.1 | 3, 4 | 306 | ||
XM_005254668.3 | XP_005254725.1 | 3, 4 | 356 | |||
XM_017022571.1 | XP_016878060.1 | 4, 5 | 390 | |||
XM_017022572.1 | XP_016878061.1 | 4, 5 | 695 | |||
XM_017022573.1 | XP_016878062.1 | 3, 4 | 516 | |||
XM_017022574.1 | XP_016878063.1 | 4, 5 | 369 | |||
XM_017022575.1 | XP_016878064.1 | 3, 4 | 309 | |||
GenBank mRNA | AK026210.1 | - | 2, 3 | 125 | ||
AK293021.1 | - | 3, 4 | 243 | |||
AK293695.1 | - | 2, 3 | 175 | |||
AK297904.1 | - | 3, 4 | 271 | |||
AL136908.1 | - | 3, 4 | 232 | |||
AL833631.1 | - | 3, 4 | 290 | |||
BC026234.1 | - | 3, 4 | 198 | 4693115 | ||
BC111000.1 | - | 3, 4 | 267 | 5751934 |
Gene Symbol: KATNBL1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003954.1 | 1 | 262 | |||
GenBank mRNA | BC067801.1 | - | 1 | 262 |