Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024713.2 | NP_078989.1 | 9, 10 | 1046 | ||
XM_017022571.1 | XP_016878060.1 | 10, 11 | 1130 | |||
XM_017022572.1 | XP_016878061.1 | 10, 11 | 1435 | |||
XM_017022573.1 | XP_016878062.1 | 9, 10 | 1256 | |||
XM_017022574.1 | XP_016878063.1 | 10, 11 | 1109 | |||
XM_017022575.1 | XP_016878064.1 | 6, 7 | 818 | |||
GenBank mRNA | AK026210.1 | - | 9, 10 | 865 | ||
AK293021.1 | - | 9, 10 | 983 | |||
AL136908.1 | - | 9, 10 | 972 | |||
AL833631.1 | - | 9, 10 | 1030 | |||
BC026234.1 | - | 9, 10 | 938 | 4693115 | ||
BC111000.1 | - | 9, 10 | 1007 | 5751934 | ||
BX389537.2 | - | 3, 4 | 223 |
Gene Symbol: KATNBL1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003954.1 | 1 | 1001 | |||
GenBank mRNA | BC067801.1 | - | 1 | 1001 |