Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002796.3 | NP_001002796.1 | 8, 9 | 1075 | ||
NM_024717.5 | NP_078993.4 | 8, 9 | 1354 | |||
XM_005272082.4 | XP_005272139.1 | 9, 10 | 1481 | |||
XM_011543649.2 | XP_011541951.1 | 9, 10 | 1555 | |||
XM_011543650.2 | XP_011541952.1 | 7, 8 | 532 | |||
XM_017009855.1 | XP_016865344.1 | 7, 8 | 755 | |||
XM_017009856.1 | XP_016865345.1 | 8, 9 | 854 | |||
XM_017009859.1 | XP_016865348.1 | 8, 9 | 679 | |||
GenBank mRNA | AK058012.1 | - | 7, 8 | 621 | ||
AY656715.1 | - | 8, 9 | 1354 | |||
AY656716.1 | - | 8, 9 | 691 | |||
BC030005.2 | - | 8, 9 | 836 | 3834212 |