Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286732.1 | NP_001273661.1 | 7 | 967 | ||
NM_024719.3 | NP_078995.2 | 7 | 967 | |||
XM_005268333.3 | XP_005268390.1 | 6 | 804 | |||
XM_017020755.1 | XP_016876244.1 | 7 | 925 | |||
XM_017020756.1 | XP_016876245.1 | 8 | 1105 | |||
GenBank mRNA | AB449887.1 | - | 7 | 870 | ||
AK026127.1 | - | 5 | 610 | |||
AK300897.1 | - | 7 | 917 | |||
AW243250.1 | - | 2 | 464 | |||
BC033071.2 | - | 5 | 691 | |||
BC062461.1 | - | 6 | 815 | |||
BC080640.1 | - | 7 | 924 | |||
BC112123.1 | - | 5 | 644 | 8327591 | ||
BC112125.1 | - | 5 | 656 | 8327593 | ||
BC143682.1 | - | 4 | 716 | |||
CB242314.1 | - | 2 | 453 |