Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006304.1 | NP_006295.1 | 1 | 153 | ||
GenBank mRNA | AK291070.1 | - | 1 | 133 | ||
AK309241.1 | - | 1 | 111 | |||
BC032782.1 | - | 1 | 113 | 5166837 | ||
CR456887.1 | - | 1 | 25 | |||
U41515.1 | - | 1 | 153 |