Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031720.3 | NP_001026890.2 | 4 | 1122 | ||
NM_024751.3 | NP_079027.2 | 4 | 1043 | |||
XM_005263222.3 | XP_005263279.1 | 4 | 1310 | |||
XM_011532248.2 | XP_011530550.1 | 4 | 1266 | |||
XM_011532249.2 | XP_011530551.1 | 4 | 1128 | |||
XM_011532252.2 | XP_011530554.1 | 4 | 1500 | |||
GenBank mRNA | AK023335.1 | - | 4 | 1301 | ||
AK225563.1 | - | 4 | 1301 | |||
AK292355.1 | - | 4 | 1125 | |||
AK303547.1 | - | 2 | 389 | |||
BC032942.2 | - | 4 | 1119 | 4838672 | ||
BX648355.1 | - | 4 | 1044 |