Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321635.1 | NP_001308564.1 | 4 | 671 | ||
NM_001321636.1 | NP_001308565.1 | 4 | 671 | |||
NM_024759.2 | NP_079035.1 | 4 | 671 | |||
NR_135745.1 | 4 | 671 | ||||
NR_135746.1 | 4 | 671 | ||||
NR_135747.1 | 4 | 671 | ||||
XM_011517302.2 | XP_011515604.1 | 4 | 1705 | |||
GenBank mRNA | AK024017.1 | - | 4 | 671 | ||
AK302414.1 | - | 2 | 186 | |||
AW082976.1 | - | 4 | 459 | |||
BC132687.1 | - | 4 | 614 | 40146709 | ||
BC144055.1 | - | 4 | 614 | 9052575 | ||
BP346328.1 | - | 3 | 261 | |||
DA184376.1 | - | 4 | 350 | |||
DC402113.1 | - | 2 | 187 |