Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010000.2 | NP_001010000.1 | 11, 12 | 1326 | ||
XM_005258144.1 | XP_005258201.2 | 12, 13 | 1648 | |||
XM_005258145.1 | XP_005258202.2 | 12, 13 | 1648 | |||
XM_005258146.2 | XP_005258203.1 | 11, 12 | 1570 | |||
GenBank mRNA | AB037735.1 | - | 12, 13 | 4599 | ||
AK001174.1 | - | 6, 7 | 655 | |||
AK289791.1 | - | 5, 6 | 586 | |||
AK302026.1 | - | 11, 12 | 1235 | |||
AK308259.1 | - | 11, 12 | 1274 | |||
BC033668.1 | - | 5, 6 | 545 | |||
BC065274.1 | - | 10, 11 | 1087 | 6164396 | ||
BX648684.1 | - | 11, 12 | 1284 |