Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024769.3 | NP_079045.1 | 4 | 806 | ||
XM_011542998.2 | XP_011541300.1 | 4 | 774 | |||
GenBank mRNA | AK026068.1 | - | 4 | 782 | ||
AY326422.1 | - | 4 | 630 | |||
AY358340.1 | - | 4 | 630 | |||
BC009371.2 | - | 4 | 452 | 4122648 |