Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031855.2 | NP_001027026.1 | 10 | 2203 | ||
NM_001289109.1 | NP_001276038.1 | 11 | 1989 | |||
NM_024778.5 | NP_079054.3 | 9 | 2080 | |||
NR_110311.1 | 10 | 2362 | ||||
XM_005262476.2 | XP_005262533.1 | 11 | 2359 | |||
GenBank mRNA | AK091777.1 | - | 10 | 2358 | ||
AK290198.1 | - | 9 | 2076 | |||
BC099847.1 | - | 10 | 2067 | 40007729 | ||
BC100671.1 | - | 10 | 2067 | 40007726 | ||
BC103491.1 | - | 9 | 1944 | 40007723 |