Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320210.1 | NP_001307139.1 | 11, 12 | 1333 | ||
NM_001321238.1 | NP_001308167.1 | 9, 10 | 1197 | |||
NM_001321239.1 | NP_001308168.1 | 9, 10 | 2047 | |||
NM_024818.4 | NP_079094.1 | 11, 12 | 2183 | |||
NM_198329.3 | NP_938143.1 | 11, 12 | 1206 | |||
GenBank mRNA | AB154406.1 | - | 11, 12 | 1142 | ||
AK026904.1 | - | 11, 12 | 1376 | |||
AK027563.1 | - | 10, 11 | 975 | |||
AK056238.1 | - | 12 | 2185 | |||
AK129616.1 | - | 12, 13 | 1242 | |||
AL136757.1 | - | 11, 12 | 1196 | |||
AL832669.1 | - | 7, 8 | 2569 | |||
AY253672.1 | - | 11, 12 | 1335 | |||
BC009737.1 | - | 11, 12 | 1312 | 3879061 | ||
BC048015.1 | - | 11, 12 | 1290 |