Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146254.1 | NP_001139726.1 | 7, 8 | 1039 | ||
NM_001146255.1 | NP_001139727.1 | 5, 6 | 772 | |||
NM_024825.3 | NP_079101.3 | 7, 8 | 1063 | |||
XM_006722901.1 | XP_006722964.1 | 7, 8 | 996 | |||
XM_006722902.1 | XP_006722965.1 | 6, 7 | 861 | |||
XM_006722903.1 | XP_006722966.1 | 5, 6 | 729 | |||
XM_011528308.1 | XP_011526610.1 | 7, 8 | 1030 | |||
XM_011528309.2 | XP_011526611.1 | 6, 7 | 955 | |||
XM_011528310.2 | XP_011526612.1 | 6, 7 | 918 | |||
XM_011528311.1 | XP_011526613.1 | 6, 7 | 897 | |||
XM_011528313.1 | XP_011526615.1 | 6, 7 | 965 | |||
XM_011528314.1 | XP_011526616.1 | 6, 7 | 886 | |||
XM_011528316.1 | XP_011526618.1 | 4, 5 | 698 | |||
XM_017027313.1 | XP_016882802.1 | 6, 7 | 920 | |||
GenBank mRNA | AK027100.1 | - | 5, 6 | 763 | ||
AK290731.1 | - | 7, 8 | 996 | |||
AK296101.1 | - | 5, 6 | 759 | |||
AK298489.1 | - | 7, 8 | 1039 | |||
BC057786.1 | - | 7, 8 | 1063 | 30347370 |