Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199120.1 | NP_001186049.1 | 5 | 480 | ||
NM_001199121.1 | NP_001186050.1 | 5 | 463 | |||
NM_024839.2 | NP_079115.1 | 5 | 456 | |||
GenBank mRNA | AF212152.2 | - | 5 | 426 | ||
AJ504713.1 | - | 5 | 416 | |||
AJ504714.1 | - | 4 | 495 | |||
AJ504715.1 | - | 4 | 485 | |||
AJ504716.1 | - | 5 | 423 | |||
AK026291.1 | - | 5 | 425 | |||
BC011730.2 | - | 5 | 418 | 4422871 | ||
BI598757.1 | - | 5 | 483 | |||
DA270919.1 | - | 5 | 456 |
Gene Symbol: TRIM39-RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199119.1 | NP_001186048.1 | 10 | 1467 | ||
GenBank mRNA | AI805914.1 | - | 1 | 106 | ||
EU012025.1 | - | 10 | 1196 |