Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024854.3 | NP_079130.2 | 12 | 1568 | ||
XM_017019976.1 | XP_016875465.1 | 6 | 882 | |||
GenBank mRNA | AK025681.1 | - | 12 | 1471 | ||
AK125461.1 | - | 12 | 1981 | |||
AK307710.1 | - | 11 | 1393 | |||
AL832441.1 | - | 12 | 1495 | |||
BC021662.1 | - | 12 | 1469 | 3826938 |