Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 16 | 2336 | ||
XM_005248376.3 | XP_005248433.1 | 16 | 2441 | |||
XM_005248377.3 | XP_005248434.1 | 16 | 2426 | |||
XM_005248378.3 | XP_005248435.1 | 16 | 2430 | |||
XM_011514135.2 | XP_011512437.1 | 16 | 2446 | |||
XM_011514136.2 | XP_011512438.1 | 16 | 2445 | |||
XM_011514137.2 | XP_011512439.1 | 16 | 2430 | |||
XM_011514138.2 | XP_011512440.1 | 16 | 2446 | |||
XM_011514139.2 | XP_011512441.1 | 16 | 2445 | |||
XM_011514140.2 | XP_011512442.1 | 15 | 2180 | |||
XM_011514141.2 | XP_011512443.1 | 17 | 2644 | |||
XM_011514142.1 | XP_011512444.1 | 16 | 2441 | |||
XM_017009880.1 | XP_016865369.1 | 15 | 2262 | |||
XM_017009881.1 | XP_016865370.1 | 16 | 3300 | |||
XM_017009882.1 | XP_016865371.1 | 16 | 2434 | |||
GenBank mRNA | AB051557.1 | - | 6 | 677 | ||
AK094564.1 | - | 5 | 358 | |||
AK097547.1 | - | 7 | 1077 | |||
AL832607.1 | - | 8 | 3947 | |||
EF553528.1 | - | 16 | 2271 |