Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 25 | 3694 | ||
XM_005248376.3 | XP_005248433.1 | 25 | 3799 | |||
XM_005248377.3 | XP_005248434.1 | 25 | 3784 | |||
XM_011514135.2 | XP_011512437.1 | 25 | 3804 | |||
XM_011514136.2 | XP_011512438.1 | 25 | 3803 | |||
XM_011514137.2 | XP_011512439.1 | 25 | 3788 | |||
XM_011514138.2 | XP_011512440.1 | 25 | 3780 | |||
XM_011514139.2 | XP_011512441.1 | 24 | 3728 | |||
XM_011514140.2 | XP_011512442.1 | 24 | 3538 | |||
XM_011514141.2 | XP_011512443.1 | 26 | 4002 | |||
XM_011514142.1 | XP_011512444.1 | 25 | 3775 | |||
XM_017009880.1 | XP_016865369.1 | 24 | 3620 | |||
XM_017009881.1 | XP_016865370.1 | 25 | 4658 | |||
GenBank mRNA | AB051557.1 | - | 15 | 2035 | ||
AK027230.1 | - | 2 | 140 | |||
AK225586.1 | - | 2 | 140 | |||
AK225607.1 | - | 2 | 140 |