Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142555.2 | NP_001136027.1 | 12, 13 | 1150 | ||
NM_024876.3 | NP_079152.3 | 13, 14 | 1518 | |||
GenBank mRNA | AF052122.1 | - | 2, 3 | 112 | ||
AK057199.1 | - | 3, 4 | 1331 | |||
BC013114.1 | - | 13, 14 | 1328 | 4342891 | ||
BC027473.1 | - | 12, 13 | 1117 | 4997823 |