Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DHDDS SIRNA Assays ›
Gene Symbol:
DHDDS
Gene Name:
dehydrodolichyl diphosphate synthase subunit
Gene Aliases:
CIT, CPT, DS, HDS, RP59
Chromosome Location:
Chr.1: 26432282 - 26471306 on Build GRCh38
UniGene ID:
Hs.369385
Species:
Human
siRNA ID
HSS188292
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DHDDS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243564.1
NP_001230493.1
6
643
NM_001243565.1
NP_001230494.1
6
628
NM_001319959.1
NP_001306888.1
7
768
NM_024887.3
NP_079163.2
7
745
NM_205861.2
NP_995583.1
7
745
XM_006710912.2
XP_006710975.1
7
804
XM_006710913.2
XP_006710976.1
7
683
XM_006710914.2
XP_006710977.1
7
680
XM_006710915.2
XP_006710978.1
6
615
XM_006710916.2
XP_006710979.1
7
794
XM_006710917.2
XP_006710980.1
7
747
XM_006710918.2
XP_006710981.1
7
703
XM_011542183.2
XP_011540485.1
7
780
XM_011542184.2
XP_011540486.1
7
801
XM_011542185.2
XP_011540487.1
6
699
XM_011542186.2
XP_011540488.1
6
699
XM_017002379.1
XP_016857868.1
7
683
XM_017002380.1
XP_016857869.1
7
780
XM_017002381.1
XP_016857870.1
7
794
XM_017002382.1
XP_016857871.1
7
703
XM_017002383.1
XP_016857872.1
6
692
XM_017002384.1
XP_016857873.1
6
645
GenBank mRNA
AB090852.1
-
6
606
AK023164.1
-
7
675
AK297134.1
-
5
563
AK308187.1
-
7
687
AK316485.1
-
6
586
BC003643.2
-
6
650
3532466
BC004117.2
-
6
570
3843062
BX648507.1
-
7
770
GU727641.1
-
6
650
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
dolichyl diphosphate biosynthetic process
polyprenyltransferase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline