Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261392.1 | NP_001248321.1 | 19 | 1975 | ||
NM_001261394.1 | NP_001248323.1 | 19 | 1903 | |||
NM_024894.3 | NP_079170.2 | 20 | 2053 | |||
NR_048552.1 | 19 | 2007 | ||||
XM_011510398.2 | XP_011508700.1 | 19 | 1920 | |||
XM_011510399.2 | XP_011508701.1 | 18 | 1858 | |||
XM_017004990.1 | XP_016860479.1 | 16 | 1578 | |||
XM_017004991.1 | XP_016860480.1 | 9 | 1030 | |||
GenBank mRNA | AK024137.1 | - | 19 | 1888 | ||
AK290680.1 | - | 20 | 2017 | |||
AK297169.1 | - | 19 | 1963 | |||
AK299617.1 | - | 19 | 1995 | |||
AL529108.3 | - | 1 | 75 | |||
BC005125.2 | - | 20 | 2033 | 3841643 | ||
DN602355.1 | - | 5 | 681 |