Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105533.1 | NP_001099003.1 | 7 | 1237 | ||
NM_024913.4 | NP_079189.4 | 8 | 1453 | |||
XM_017012649.1 | XP_016868138.1 | 7 | 1929 | |||
GenBank mRNA | AK289978.1 | - | 8 | 1450 | ||
AL832619.2 | - | 8 | 1408 | |||
AL832638.2 | - | 8 | 1277 | |||
BC017764.1 | - | 4 | 450 | |||
BC018685.1 | - | 7 | 988 |