Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105533.1 | NP_001099003.1 | 3 | 718 | ||
NM_024913.4 | NP_079189.4 | 4 | 934 | |||
XM_017012649.1 | XP_016868138.1 | 3 | 1410 | |||
GenBank mRNA | AK055624.1 | - | 3 | 1058 | ||
AK289978.1 | - | 4 | 931 | |||
AL832619.2 | - | 4 | 889 | |||
AL832638.2 | - | 4 | 758 | |||
AY358220.1 | - | 3 | 510 | |||
AY780785.1 | - | 3 | 487 | |||
BC018685.1 | - | 3 | 469 | |||
BC046360.1 | - | 3 | 934 |