Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122841.2 | NP_001116313.1 | 2 | 184 | ||
NM_024919.4 | NP_079195.3 | 2 | 334 | |||
NR_110312.1 | 3 | 414 | ||||
XM_011536137.1 | XP_011534439.1 | 3 | 414 | |||
XM_011536138.1 | XP_011534440.1 | 3 | 230 | |||
XM_011536140.1 | XP_011534442.1 | 2 | 269 | |||
XM_011536145.1 | XP_011534447.1 | 2 | 184 | |||
XM_017011317.1 | XP_016866806.1 | 3 | 414 | |||
GenBank mRNA | AK074110.1 | - | 3 | 271 | ||
AK097179.1 | - | 2 | 334 | |||
AK097579.1 | - | 2 | 184 | |||
AK125963.1 | - | 3 | 386 | |||
BC137144.1 | - | 2 | 292 | 9021141 | ||
DB019858.1 | - | 3 | 414 |