Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303228.1 | NP_001290157.1 | 3 | 650 | ||
NM_024943.2 | NP_079219.1 | 3 | 650 | |||
XM_005262670.3 | XP_005262727.1 | 4 | 700 | |||
XM_011513749.2 | XP_011512051.1 | 4 | 697 | |||
XM_011513750.2 | XP_011512052.1 | 4 | 698 | |||
XM_011513751.2 | XP_011512053.1 | 4 | 701 | |||
XM_011513752.2 | XP_011512054.1 | 4 | 701 | |||
XM_011513753.2 | XP_011512055.1 | 4 | 615 | |||
XM_011513754.2 | XP_011512056.1 | 3 | 913 | |||
XM_017008629.1 | XP_016864118.1 | 4 | 696 | |||
GenBank mRNA | AK026888.1 | - | 3 | 650 | ||
BC030803.1 | - | 3 | 591 | 4806992 | ||
BC060512.1 | - | 3 | 616 |