Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308116.1 | NP_001295045.1 | 8 | 1496 | ||
NM_024947.3 | NP_079223.3 | 8 | 1481 | |||
XM_005247789.2 | XP_005247846.1 | 8 | 1511 | |||
XM_005247791.3 | XP_005247848.1 | 8 | 1511 | |||
XM_005247793.2 | XP_005247850.1 | 8 | 1511 | |||
XM_006713753.1 | XP_006713816.1 | 8 | 1480 | |||
XM_006713754.3 | XP_006713817.1 | 8 | 1602 | |||
XM_006713756.2 | XP_006713819.1 | 6 | 1253 | |||
XM_006713757.3 | XP_006713820.1 | 8 | 1511 | |||
XM_011513186.2 | XP_011511488.1 | 7 | 1412 | |||
XM_011513187.2 | XP_011511489.1 | 7 | 1412 | |||
XM_011513188.2 | XP_011511490.1 | 7 | 1352 | |||
XM_011513189.2 | XP_011511491.1 | 7 | 1352 | |||
XM_017007235.1 | XP_016862724.1 | 8 | 1499 | |||
XM_017007236.1 | XP_016862725.1 | 8 | 1487 | |||
XM_017007237.1 | XP_016862726.1 | 7 | 1352 | |||
XM_017007238.1 | XP_016862727.1 | 6 | 1253 | |||
XM_017007239.1 | XP_016862728.1 | 5 | 1052 | |||
GenBank mRNA | AF380154.1 | - | 8 | 1418 | ||
AF444193.1 | - | 7 | 1433 | |||
AJ320486.3 | - | 8 | 1475 | |||
AK304411.1 | - | 7 | 1322 | |||
BC131772.1 | - | 8 | 1460 | 40123356 | ||
EF560717.1 | - | 8 | 1456 |