Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004342.6 | NP_004333.1 | 12 | 1943 | ||
NM_033138.3 | NP_149129.2 | 13 | 2708 | |||
NM_033139.3 | NP_149130.1 | 11 | 1758 | |||
NM_033140.3 | NP_149131.1 | 10 | 1680 | |||
NM_033157.3 | NP_149347.2 | 13 | 2018 | |||
XM_017012650.1 | XP_016868139.1 | 11 | 2240 | |||
XM_017012651.1 | XP_016868140.1 | 11 | 1556 | |||
XM_017012652.1 | XP_016868141.1 | 11 | 1717 | |||
XM_017012653.1 | XP_016868142.1 | 10 | 1475 | |||
XM_017012654.1 | XP_016868143.1 | 10 | 1677 | |||
GenBank mRNA | AF247820.3 | - | 11 | 2127 | ||
AF315788.1 | - | 1 | 674 | |||
AK289686.1 | - | 13 | 2011 | |||
AK298524.1 | - | 12 | 1569 | |||
AK300282.1 | - | 10 | 1649 | |||
AK304729.1 | - | 12 | 1490 | |||
BC040354.1 | - | 12 | 1678 | 4753285 | ||
BQ010124.1 | - | 3 | 669 | |||
BX538339.1 | - | 2 | 182 | |||
BX648808.1 | - | 10 | 1638 | |||
BX649088.1 | - | 2 | 121 | |||
M64110.1 | - | 11 | 1588 | |||
M83216.1 | - | 13 | 2471 |