Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006981.3 | NP_008912.2 | 5 | 1846 | ||
NM_173199.2 | NP_775291.1 | 5 | 1846 | |||
NM_173200.2 | NP_775292.1 | 6 | 1918 | |||
XM_017015162.1 | XP_016870651.1 | 4 | 1778 | |||
GenBank mRNA | AK226108.1 | - | 5 | 1828 | ||
BX649089.1 | - | 2 | 214 | |||
D78579.1 | - | 5 | 1848 | |||
D85242.1 | - | 3 | 217 | |||
S81243.1 | - | 5 | 1733 | |||
U12767.1 | - | 4 | 1209 | |||
X89894.1 | - | 5 | 1804 |