Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025074.6 | NP_079350.5 | 62 | 9913 | ||
XM_006714316.2 | XP_006714379.1 | 61 | 9932 | |||
GenBank mRNA | AB040933.2 | - | 24 | 4158 | ||
AK092082.1 | - | 6 | 1076 | |||
AL833581.1 | - | 2 | 243 | |||
BX647420.1 | - | 7 | 1307 | |||
BX647949.1 | - | 12 | 2193 |